Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thiol resin
Thiol resin MP, fine beads 100-200 mesh Loading: 2.4-2.5 mmol/g
Thiol resin MP, fine beads 100-200 mesh Loading: 2.4-2.5 mmol/g
Thiol resin MP, fine beads 100-200 mesh Loading: 2.4-2.5 mmol/g
Supplier:
Supra Sciences
Certificates
Expand image
Thiol resin MP, fine beads 100-200 mesh Loading: 2.4-2.5 mmol/g
Supplier:
Supra Sciences
About this item
:
Specification Test Results
Colour:
Off-white
Form:
Granule
Loading:
2.4-2.5 mmol/g
Particle size:
100-200 mesh
Specification Test Results
Related Information
Documentation
SDS
ghs_SPMP16
Datasheet
Thiol
Documentation