$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Rink Amide resin
Rink Amide resin on polystyrene (PS), beads 100-200 mesh Loading: 0.6-0.7 mmol/g
Rink Amide resin on polystyrene (PS), beads 100-200 mesh Loading: 0.6-0.7 mmol/g
:
Supra Sciences
Rink Amide resin on polystyrene (PS), beads 100-200 mesh Loading: 0.6-0.7 mmol/g
:
Supra Sciences
:
Colour:
Off-white
Form:
Granule
Loading:
0.6-0.7 mmol/g
Particle size:
100-200 mesh
ghs_SPPS38
Rink