Morpholine resin (MP), beads, macroporous 330-1225 µm, 15-50 mesh (>90% within), polymer bound, equivalent of N-methylmorpholine (NMM), utilized as an acid scavenger Loading: 3.4-3.6 mmol/g
Supplier: Supra Sciences
Certificates
About this item
:
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:3.4-3.6 mmol/g
- Particle size:330-1225 μm (15-50 mesh)