Merrifield Resin, beads 100-200 mesh Loading: 1.0-1.2 mmol/g
Supplier: Supra Sciences
Certificates
About this item
CAS Number:70024-51-0
:
MDL Number:MFCD00146406
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:1.0-1.2 mmol/g
- Particle size:100-200 mesh