Diethanolamine on resin, fine beads 100-200 mesh Loading: 1.2-1.4 mmol/g
Supplier: Supra Sciences
Certificates
About this item
CAS Number:111-42-2
:
Formula:C₄H₁₁NO₂
MDL Number:MFCD00002843
:105.14 g/mol
Specification Test Results
- Colour:Off-white
- Form:Granule
- Loading:1.2-1.4 mmol/g
- Particle size:100-200 mesh