ContactUsLinkComponent
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
Catalog # 75832-478
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
CRITERION™ Dehydrated Culture Media, Barney Miller Broth with Indicator, Hardy Diagnostics
Catalog # 75832-478
Supplier:  Hardy Diagnostics
Supplier Number:  C8992
Product available on GSA Advantage®
CAS Number:  
Specifications
  • Description:
    Barney Miller Broth with Indicator
  • Size:
    2 kg
  • Cat. No.:
    75832-478
  • Supplier no.:
    C8992
Specifications
About this item
Hardy Diagnostics CRITERION™ Barney Miller Broth with Indicator is recommended for the isolation and detection of beer spoilage microorganisms, specifically lactic acid bacteria
  • Wide mouth opening reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip enables 'no slip' finger holds
  • Stackable containers for efficient storage
  • Increased solubility; dissolves quickly
  • Custom and bulk products available to meet customer specifications
  • Complete technical/instruction sheets, available in HUGO software
  • Unparalleled performance; tested for optimal growth
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards. This broth is supplied in a 2kg bucket with convenient screw lid.
Indicator allows for early detection of growth by yellow color change due to pH shift.