Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Desmin Rabbit Monoclonal Antibody [clone: Clone: RM234]
Anti-Desmin Rabbit Monoclonal Antibody [clone: Clone: RM234]
Catalog # 103255-608
Supplier:  RevMAb Biosciences
undefined
Anti-Desmin Rabbit Monoclonal Antibody [clone: Clone: RM234]
Catalog # 103255-608
Supplier:  RevMAb Biosciences
Supplier Number:  31-1124-00

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Desmin
  • Antigen Symbol:
    Desmin
  • Clonality:
    Monoclonal
  • Clone:
    Clone: RM234
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Bovine,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Format:
    Purified
  • Form:
    Liquid
  • Gene ID:
    1674
  • Storage Buffer:
    50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
  • Storage Temperature:
    -20.0°C
  • Shipping Temperature:
    4°C
  • Immunogen:
    A peptide corresponding to human Desmin
  • Purification:
    Protein A affinity purified from an animal origin-free culture supernatant
  • Cat. No.:
    103255-608
  • Supplier no.:
    31-1124-00

Specifications

About this item

This antibody reacts to human and mouse Desmin. This antibody may also react to rat and Bovine Desmin, as predicted by immunogen homology.

Immunohistochemistry (IHC): 1:1000-1:4000 dilution;
Western Blot (WB): 1:1000-1:2000 dilution.