Anti-BRAF V600E Rabbit Monoclonal Antibody [clone: RM8]
Catalog # 103255-598
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:BRAF V600E Mutant
- Antigen Symbol:BRAF V600E
- Clonality:Monoclonal
- Clone:Clone: RM8
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Epitope:V600E point mutation in BRAF
- Format:Purified
- Form:Liquid
- Gene ID:673
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:A peptide corresponding to BRAF V600E mutant
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-598
- Supplier no.:31-1042-00
Specifications
About this item
This antibody reacts to the BRAF V600E mutant. No cross reactivity with wild type BRAF.
Immunohistochemistry (IHC): 2 – 10 ug/mL;
Immunocytochemistry (ICC): 2 – 10 ug/mL;
Western Blot (WB): 0.5 – 2 ug/mL