Anti-H2BK20ac Rabbit Monoclonal Antibody [clone: RM235]
Catalog # 103255-302
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:Acetyl-Histone H2B (Lys20), H2BK20ac
- Antigen Symbol:H2BK20ac
- Clonality:Monoclonal
- Clone:Clone: RM235
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Epitope:Acetyllysine 20 in Histone H2B
- Format:Purified
- Form:Liquid
- Gene ID:3018
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:An acetyl-peptide corresponding to Acetyl-Histone H2B (Lys20).
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-302
- Supplier no.:31-1113-00
Specifications
About this item
This antibody reacts to Histone H2B acetylated at Lysine 20 (K20ac). No cross reactivity with non-modified Lysine 20 or other acetylated Lysines in histone H2B.
Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 0.5 ug/mL.