Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-H2AZK7ac Rabbit Monoclonal Antibody [clone: RM222]
Anti-H2AZK7ac Rabbit Monoclonal Antibody [clone: RM222]
Catalog # 103255-524
Supplier:  RevMAb Biosciences
undefined
Anti-H2AZK7ac Rabbit Monoclonal Antibody [clone: RM222]
Catalog # 103255-524
Supplier:  RevMAb Biosciences
Supplier Number:  31-1102-00

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Acetylated Histone H2AZ Lysine 7, H2AZK7ac
  • Antigen Symbol:
    H2AZK7ac
  • Clonality:
    Monoclonal
  • Clone:
    Clone: RM222
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Multiple
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    Acetyllysine 7 in Histone H2AZ
  • Format:
    Purified
  • Form:
    Liquid
  • Gene ID:
    3015
  • Storage Buffer:
    50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
  • Storage Temperature:
    -20.0°C
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4°C
  • Immunogen:
    An acetyl-peptide corresponding to Acetyl-Histone H2A.Z (Lys7)
  • Purification:
    Protein A affinity purified from an animal origin-free culture supernatant
  • Cat. No.:
    103255-524
  • Supplier no.:
    31-1102-00

Specifications

About this item

This antibody reacts to Histone H2A.Z acetylated at Lysine 7 (K7ac). No cross reactivity with non-modified Lysine 7 or other acetylated Lysines in histone H2A.

Western Blot: 0.5 ug/mL – 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.05 ug/mL – 0.5 ug/mL.