Anti-H4K20ac Rabbit Monoclonal Antibody [clone: RM205]
Catalog # 103255-498
Supplier: RevMAb Biosciences
Specifications
- Antibody Type:Primary
- Antigen Name:Acetylated Histone H4 Lysine 20, H4K20ac
- Antigen Symbol:H4K20ac
- Clonality:Monoclonal
- Clone:Clone: RM205
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Multiple
- Western Blot:Yes
- Size:100 µg
- Epitope:Acetyllysine 20 in Histone H4
- Format:Purified
- Form:Liquid
- Gene ID:121504
- Storage Buffer:50% Glycerol/PBS with 1% BSA and 0.09% sodium azide
- Storage Temperature:-20.0°C
- Concentration:1 mg/mL
- Shipping Temperature:4°C
- Immunogen:An acetyl-peptide corresponding to the Acetyl-Histone H4 (Lys20).
- Purification:Protein A affinity purified from an animal origin-free culture supernatant
- Cat. No.:103255-498
- Supplier no.:31-1084-00
Specifications
About this item
This antibody reacts to Histone H4 acetylated at Lysine 20 (K20ac). No cross reactivity with other acetylated Lysines in Histone H4.
Western Blot: 1 ug/mL - 2 ug/mL;
ELISA: 0.2 ug/mL - 1 ug/mL;
Multiplex: 0.1 ug/mL – 1 ug/mL.