Test Kits, Sulfite , LaMotte®
Catalog # 66170-510
Supplier: LaMotte
Small Business Enterprise
CAS Number:
Specifications
- Description:Dropper Pipet
- Cat. No.:66170-510
- Supplier no.:7175
- No. of tests:50 at 100 ppm
Specifications
About this item
An iodide-iodate titrant oxidizes sulfite to sulfate under acid conditions, until all of the sulfite is reacted
The titrant then reacts with starch to form a blue color signifying the endpoint.