To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:3-HB R2 Set (Autokit)
- Size:2 x 9 mL
- Cat. No.:11001-662
- Storage temperature:+2…+10 °C
- Supplier no.:413-73601
Specifications
About this item
The Autokit 3-Hb R2 Set is an in vitro assay for the quantitative determination of 3-hydroxybutyrate (3-HB) in serum or plasma.
- Utilizes cyclic enzymatic reactions
- High sensitivity and high specificity
- Quantitative determination of 3-HB in serum or plasma
The Autokit 3-Hb R2 Set is an in vitro assay for the quantitative determination of 3-hydroxybutyrate (3-HB) in serum or plasma. It utilizes cyclic enzymatic reactions for high sensitivity and high specificity.