ContactUsLinkComponent
Pierce Imject™ Purification Buffer Salts, Thermo Scientific
Pierce Imject™ Purification Buffer Salts, Thermo Scientific
  PI77159
 :  
Pierce Imject™ Purification Buffer Salts, Thermo Scientific
  PI77159
 :  77159
 :  
  • Size:
    5 g
  • Cat. No.:
    PI77159
  • pH:
    7.2
  • Supplier no.:
    77159
Imject™ purification buffer salts is a dry powder for preparing 60 mL of solution for buffer-exchange of immunogens following conjugation.
 : For research use only. Not for use in diagnostic procedures.