Autokit Total Ketone Bodies R2 Set, FUJIFILM Irvine Scientific
Catalog # 11001-656
Supplier: Wako Life Sciences
CAS Number:
Specifications
- Description:Total Ketone Bodies R2 Set (Autokit)
- Size:2 x 9 mL
- Cat. No.:11001-656
- Storage temperature:+2…+10 °C
- Supplier no.:411-73401
Specifications
About this item
The Autokit Total Ketone Bodies R2 Set is an in vitro assay for the quantitative determination of total ketone bodies in serum or plasma.
- For research use only
- Quantitative determination of total ketone bodies
- In vitro assay
The Autokit Total Ketone Bodies R2 Set is an in vitro assay for the quantitative determination of total ketone bodies [acetoacetone (AcAc) + 3 hydroxybutyrate (3-HB)] in serum or plasma. For Research Use Only. Contents: Enzyme 2 x for 9 mL, Diluent 2 x for 9 mL.