Autokit Total Ketone Bodies R1 Set, FUJIFILM Irvine Scientific
11001-654
:
- Description:Total Ketone Bodies R1 Set (Autokit)
- Size:2 x 27 mL
- Cat. No.:11001-654
- Storage temperature:+2…+10 °C
- Supplier no.:415-73301
The Autokit Total Ketone Bodies R1 Set is an in vitro assay for the quantitative determination of total ketone bodies in serum or plasma
- For research use only
- Quantitative determination of total ketone bodies
- In vitro assay
The Autokit Total Ketone Bodies R1 Set is an in vitro assay for the quantitative determination of total ketone bodies [acetoacetone (AcAc) + 3 hydroxybutyrate (3-HB)] in serum or plasma. For Research Use Only. Contents: Thio-NAD 2 x for 27 mL, Buffer 2 x 27 mL.