Blocks and stabilizes coated ELISA plates and other solid phase substrates.
- No Bovine serum proteins
- Provides structural support and crowding agents
- Minimizes background interactions and issues
- Eliminates BSE regulatory concerns
- ImmunO4 APMPC™ is compatible and works well in combination with ImmunO4 Stabilizers
- Room temperature stable reagent
- Stable through freeze cycles
- Superior coat blocking
APMPC™ is a superior diluent that blocks, seals, and stabilizes with a consistent coat
APMPC™ can be used for antigens or antibodies. APMPC™ can be used for blocking and as a diluent buffer. An additional blocking step may not be required, potentially enabling blocking and stabilization in one step. Coated plates dried with APMPC™ have a long term shelf life, even at room temperature, when stored in a cool and dry environment. Antimicrobial agents are included to retard bacterial growth. APMPC™ should be tested for efficacy and stability in any new assay system.
This Coat Block was developed to substantially reduce Non Specific Antibody Binding and eliminate non-specific protein interactions. It is supplied as a one component liquid, ready-to-use formula.
- Form:Liquid
- Shelf Life:24 Months at 2° to 8°C
- Storage Temperature:2° to 8°C
- Application:Immunoassay, ELISA, Blotting
- Color:Pale Yellow to Tan