Blocks and stabilizes coated ELISA plates and other solid phase substrates.
- No Mammalian proteins
- Minimizes background interactions and issues
- Eliminates BSE regulatory concerns
- ImmunO4 APNMC™ synergizes with HRPZR Super Shield™ Stabilizer to further improve unwanted non specific antibody binding
- Room temperature stable reagent
- Stable through freeze cycles
- Superior coat block
APNMC™ is a superior diluent that blocks, seals, and stabilizes with a consistent coat
APNMC™ can be used for antigens or antibodies. APNMC™ can be used for blocking and as a diluent buffer. An additional blocking step may not be required, potentially enabling blocking and stabilization in one step. Coated plates dried with APNMC™ have a long-term shelf life, even at room temperature, when stored in a cool and dry environment. Antimicrobial agents are included to retard bacterial growth. APNMC™ should be tested for efficacy and stability in any new assay system.
This Coat Block was developed to substantially reduce non-specific Antibody binding and eliminate non-specific protein interactions. It provides superior blocking and is supplied as a one component liquid, ready-to-use formula.
- Form:Liquid
- Shelf Life:24 Months at 2° to 8°C
- Storage Temperature:2° to 8°C
- Application:Immunoassay, ELISA, Blotting
- Color:Pale Yellow to Tan