Order Entry
Puerto Rico
ContactUsLinkComponent
N-Butanoyl-L-homoserine lactone ≥97%, white solid
N-Butanoyl-L-homoserine lactone ≥97%, white solid
Catalog # 102987-486
Supplier:  Adipogen
CAS Number:  
N-Butanoyl-L-homoserine lactone ≥97%, white solid
Catalog # 102987-486
Supplier:  Adipogen
Supplier Number:  CDX-B0267-M010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Vial
  • Size:
    10 mg
  • Cat. No.:
    102987-486
  • Supplier no.:
    CDXB0267M010

Specifications

About this item

 :
 :171.2 g/mol
CAS Number:67605-85-0
Formula:C₈H₁₃NO₃
MDL Number:MFCD12912262