ContactUsLinkComponent
Mohr Pinchcock Clamps, Humboldt Manufacturing
  89414-928
Mohr Pinchcock Clamps, Humboldt Manufacturing
  89414-928
 :  H-8620
  • Description:
    Pinchcock Tubing Clamp
  • Length:
    82mm
  • Cat. No.:
    89414-928
  • Supplier no.:
    H-8620
This clamp is made from one-piece, rectangular, nickel-plated brass spring wire with rounded edges
It is self aligning and has molded finger grips.