Order Entry
Puerto Rico
ContactUsLinkComponent
 
Cat Albumin (from Serum)
  103219-918
 :  
undefined
Cat Albumin (from Serum)
  103219-918
 :  FSA62-0100
 :  

 

  • Source:
    Serum
  • Species:
    Cat
  • Size:
    10 g
  • Protein/Peptide Name:
    Albumin
  • Cat. No.:
    103219-918
  • Supplier no.:
    FSA62-0100

 

 

Prepared from feline serum by heat-shock fractionation.

  • Purity: ≥ 96%
  • IgG level: not detected

Equitech-Bio offers a wide range of heat-shock and proprietary Cohn method processed animal serum albumins. With our high purity albumins, researchers can formulate and customize their own specialized media for specific applications. Prepared from feline serum by heat-shock fractionation.