Goat Plasma, Equitech- Bio, Inc.
At Equitech-Bio we maintain our own herd of goats
- Sterility tested
- Endotoxin and mycoplasma testing available
- Lot to lot consistency
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Mycoplasma testing available. Heat-inactivation is available if desired.
Worming histories are controlled and documented.
Frequently Bought Together