Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Certificates
About this item
Available raw, non-sterile filtered, or sterile filtered
- Sterility tested
- Endotoxin and mycoplasma testing available
- Lot to lot consistency
Raw plasma is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Plasma from specific breeds are available by request. Sterile-Filtered plasma is sterility tested and sterile-filtered to 0.2 micron absolute. Mycoplasma testing available. Heat-inactivation is available if desired.
Collected with sodium citrate.
Certifications: Certified U.S. origin.