Goat Serum, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Certificates
About this item
Specially formulated for use as a diluent/buffer in ELISA test kits
- Sterility tested
- Endotoxin and mycoplasma testing available
- Lot to lot consistency
Equitech-Bio’s acclaimed donor goat serum. Engineered in collaboration for the first generation HIV Assay. Worming histories are controlled and documented. Physiochemical constituents are measured to ensure lot-to-lot consistency. Ideal as a blocking agent in Western blotting. Specially formulated for use as a diluent/buffer in ELISA test kits. Useful as tissue culture media supplement. Heat inactivation available if desired, and is shown with the addition of HI after the catalog code.
Useful as tissue culture media supplement.
Specifications
- Catalog No:
- 103219-586
- 103219-584
- 103219-576
- 103219-578
- Storage Temperature:
- -20° to 5°C
- -20° to 5°C
- -20° to 5°C
- -20° to 5°C