Canine Serum, Equitech- Bio Inc.
Supplier: Equitech-Bio
Certificates
About this item
Collected from sexually mature, mixed sex, mixed breed dogs.
- Sterility tested
- Endotoxin and mycoplasma testing available
- Lot to lot consistency
Raw serum is non-sterile. Low hemoglobin levels. Physiochemical constituents measured to ensure lot-to-lot consistency. Serum from specific breeds are available by request. Sterile-Filtered serum is sterility tested and sterile-filtered to 0.2 micron absolute. Endotoxin and mycoplasma testing available. Heat-inactivation is available if desired.
Specifications
- Catalog No:
- 103219-520
- 103219-518
- 103219-040
- 103219-042
- Storage Temperature:
- -20° to 5°C
- -20° to 5°C
- -20° to 5°C
- -20° to 5°C