Order Entry
Puerto Rico
ContactUsLinkComponent
Dog Albumin (from Serum)
Dog Albumin (from Serum)
Supplier:  Equitech-Bio
Certificates
undefined
Dog Albumin (from Serum)
Supplier:  Equitech-Bio
Certificates

Specifications

  • Source:
    Serum
  • Species:
    Dog
  • Form:
    Powder
  • Protein/Peptide Name:
    Albumin

Specifications

Related Information