Neuronal Tissue Dissociation
Catalog # 10497-566
Supplier: Avantor
CAS Number:
Specifications
- Source:
- Species:
- Size:100 mg
- Environmentally Preferable:
- Enzyme Name:Neuronal Tissue Dissociation
- Cat. No.:10497-566
- Supplier no.:NM100200
Specifications
About this item
NeuroPapain™ enzyme is an optimized formulation of the enzyme papain that is ideal for enzymatic dissociation of neuronal tissues.
NeuroPapain™ enzyme provides rapid and safe enzymatic digestion of primary rodent neuronal tissues when used in conjuction with NeuroPrep Medium. Neuronal tissues are more easily dissociated via mechanical trituration and the number of viable dissociated cells can be increased by up to 100% while treating with this enzyme.