Order Entry
United States
ContactUsLinkComponent
Ethanol absolute, EMPROVE® EXPERT USP, JP, Ph. Eur., BP, ChP suitable for use as excipient, SAFC®
Ethanol absolute, EMPROVE® EXPERT USP, JP, Ph. Eur., BP, ChP suitable for use as excipient, SAFC®
Supplier:  MilliporeSigma
Certificates
Ethanol absolute, EMPROVE® EXPERT USP, JP, Ph. Eur., BP, ChP suitable for use as excipient, SAFC®
Supplier:  MilliporeSigma
Certificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (m/m):
    ≥ 99,2 %
  • Assay (V/V):
    ≥ 99,5 %
  • Identity (IR):
    conforms
  • Identity 1 (ChP):
    conforms
  • Appearance:
    conforms
  • Clarity of solution:
    conforms
  • Color of solution:
    conforms
  • Clarity and color of solution (ChP):
    conforms
  • Acidity or alkalinity:
    ≤ 30 ppm
  • Acidity or alkalinity (ChP):
    conforms
  • Density (d 20 °C/20 °C):
    0,790 - 0,.793
  • Density (d 15,56 °C/15,56 °C):
    ≤ 0,7962
  • Density (d 15 °C/15 °C):
    0,794 - 0,797
  • Specific gravity (d 15 °C/15 °C):
    0,79422 - 0,79679
  • Absorbance (at 240 nm):
    ≤ 0,40
  • Absorbance (between 250 nm and 260 nm):
    ≤ 0,30
  • Absorbance (between 270 nm and 340 nm):
    ≤ 0,10
  • The spectrum shows a steadily descending curve with no observable peaks or shoulders:
    conforms
  • Absorbance (ChP) (at 240 nm):
    ≤ 0.08
  • Absorbance (ChP) (between 250 nm and 260 nm):
    ≤ 0,06
  • Absorbance (ChP) (between 270 nm and 340 nm):
    ≤ 0,02
  • Volatile impurities (GC) (Acetaldehyde and Acetal):
    ≤ 10 ppm
  • Volatile impurities (GC) (Benzene):
    ≤ 2 ppm
  • Volatile impurities (GC) (Methanol):
    ≤ 100 ppm
  • Volatile impurities (GC) (Total of other impurities):
    ≤ 300 ppm
  • Cyclohexane (GC):
    ≤ 50 ppm
  • Evaporation residue:
    ≤ 25 mg/l
  • Non-volatile matter:
    ≤ 25 mg/l
  • Bacterial endotoxins:
    ≤ 2,5 I.U./ml

Specification Test Results

Related Information