ContactUsLinkComponent
Lens and Mirror Set
Lens and Mirror Set
Catalog # 470220-586
Supplier:  Wards
undefined
Lens and Mirror Set
Catalog # 470220-586
Supplier:  Wards
Supplier Number:  PH0201601
Specifications
  • Description:
    Lens and Mirror Set
  • Cat. No.:
    470220-586
  • Supplier no.:
    PH0201601
Specifications
About this item
Perfect compliment for a ray box.
  • Universal mirror
  • Frosted lenses
Optics lens/mirror Kit, includes 1, Acrylic Double Convex frosted lens 89 mm long, 1 Acrylic Double Concave frosted lens 86mm long, 1 Universal Stainless steel mirror - can achieve the following mirrors - Plane, concave, convex - Supplied in Plastic box.