ContactUsLinkComponent
Biox Tri-Cornered Beakers
Biox Tri-Cornered Beakers
Catalog # 75799-910
Supplier:  TWD Scientific
Supplier #: BX93-0050
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Biox Tri-Cornered Beakers
Catalog # 75799-910
Supplier:  TWD Scientific
Supplier Number:  BX93-0050
Supplier #: BX93-0050
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Log in for availability
Select an Option
50 ml (1.7 oz.)
Select an Option
Clear
Select an Option
75799-910
Select an Option
93-0050
About this item
Tri-Cornered Beakers are designed to provide three dripless pouring spouts
  • Strong medical-grade polypropylene construction
  • Equipped with three dripless pouring spouts
  • Good impact strength
  • Translucent and autoclavable
  • Disposable, easy to clean, and durable enough for reuse
  • Molded Graduations
Available in six different sizes.