Order Entry
Puerto Rico
ContactUsLinkComponent
BBL™ Hemoglobin Solution 2%, BD
BBL™ Hemoglobin Solution 2%, BD
Catalog # 90000-410
Supplier:  BD
CAS Number:  
BBL™ Hemoglobin Solution 2%, BD
Catalog # 90000-410
Supplier:  BD
Supplier Number:  211874
CAS Number:  

Specifications

  • Description:
    BBL™ Hemoglobin Solution 2%
  • Size:
    100 mL
  • Cat. No.:
    90000-410
  • Supplier no.:
    211874

Specifications

About this item

Hemoglobin solution is ready to use to enrich a variety of mediums for the cultivation of fastidious organisms.

  • Contains an essential growth factor for some Haemophilus species
  • More selective formulations of chocolate agar can be prepared by adding antimicrobial solutions
  • To inhibit the growth of unwanted organisms

This hemoglobin solution is prepared as dehydrated culture media.