Ultra Cyano HPLC Columns, Restek
Supplier: Restek
Certificates
About this item
The Ultra line represents Restek's broadest selection of stationary phases on a single silica support.
- USP L10
- High-purity
Made of high-purity, type-B silica that minimizes activity and creates high-density bonding, these columns are designed for selective and reliable HPLC applications.
The Restek® Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired. It can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes. It is an excellent choice for the analysis of protonated bases.
Specifications
- Catalog No:
- 10059-654
- 10059-650
- 10059-652
- 10059-662
- 10059-664
- 10059-658
- 97040-468
- 10059-660
- 97004-566
- 10059-666
- Description:
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- Carbon Load:
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Carbon Load: 8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH Range:
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- Temperature Limit:
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C