Ultra Silica HPLC Prep Columns, Restek
: Restek
Restek makes it easy to scale up your separations.
- Easy scale-up from Restek analytical columns
- Popular bonded phases
We offer a wide range of semipreparative and preparative-scale columns packed with many of our popular Ultra, Allure®, Pinnacle® DB, and Viva bonded phases on a 5 μm spherical silica.
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.
Other stationary phases are also available.
: We strongly recommend ordering a semi-prep or prep column only after evaluating the desired separation on an equivalent analytical-scale column. Because we cannot reuse a column or the silica it contains once it has left our facility, we cannot accept returns of large-scale columns.
- Catalog No:
- 10059-592
- 10059-594
- 10059-602
- Description:
- HPLC Prep Column
- HPLC Prep Column
- HPLC Prep Column
- Endcapped:
- No
- No
- No
- pH Range:
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- Temperature Limit:
- 80°C
- 80°C
- 80°C