Ultra Silica HPLC Columns, Restek
Supplier: Restek
Certificates
About this item
The Ultra line represents Restek's broadest selection of stationary phases on a single silica support.
- USP L3
- Base-deactivated
Made of high-purity, type-B silica that minimizes activity and creates high-density bonding, these columns are designed for selective and reliable HPLC applications.
Base-deactivated spherical silica is useful for normal-phase or HILIC separations.
Specifications
- Catalog No:
- 10059-586
- 10059-588
- 10059-590
- 10059-596
- 10059-598
- 10059-584
- 10059-600
- Description:
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- HPLC Column
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH Range:
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- Temperature Limit:
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C
- 80°C