Viva columns are based on a wide pore material we designed for optimal large-molecule separations.
- USP L7
- Excellent for separating peptides or proteins
- Rugged, spherical particles with 300 Å pore size
- High proportion of pore/surface area available to large molecules
In developing Viva silica, we found that although many commercial wide-pore silicas meet the standard 300 Å mean pore size, most have very broad distributions about this mean, with a significant portion of their pore volume falling below 150 Å.
C8 is a conventional monomeric octylsilane column offering a shorter alkyl chain to provide less hydrophobic retention and improved basic peak shape over a traditional C18 phase. Like our C18, this general-purpose Restek® C8 is suitable for a wide range of compounds from acidic through slightly basic.
This means a large portion of the surface area is unavailable to larger molecules. Viva columns have a narrow distribution around the mean pore size, permitting a larger portion of the silica surface to play a role in the separation.
- Catalog No:
- 10058-138
- 10058-134
- 10058-136
- Description:
- Viva HPLC Column
- Viva HPLC Column
- Viva HPLC Column
- Carbon Load:
- Carbon Load: 5%
- Carbon Load: 5%
- Carbon Load: 5%
- Endcapped:
- Yes
- Yes
- Yes
- pH Range:
- 2.5 to 8
- 2.5 to 8
- 2.5 to 8
- Temperature Limit:
- 80°C
- 80°C
- 80°C