Orders LinkContactUsLinkComponent
Steri-Strip™ Compound Benzoin Tincture, 3M™
Steri-Strip™ Compound Benzoin Tincture, 3M™
Benzoin Tincture, 0.6 mL
Catalog # 75788-114
Supplier:   Healthcare Products
Supplier #:  MMM C1544
Steri-Strip™ Compound Benzoin Tincture, 3M™
Catalog # 75788-114
Steri-Strip™ Compound Benzoin Tincture, 3M™
Benzoin Tincture, 0.6 mL
Price
$395.89
Log in for availability
Case of 4
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An alcohol solution of compound benzoin that may be used to increase adhesion of 3M™Steri-Strip™Skin closures to the skin.
Specifications
  • Description:
    Benzoin Tincture
  • Size:
    0.6 mL