Order Entry
Puerto Rico
ContactUsLinkComponent
Phenol Red Broth with Trehalose, Hardy Diagnostics
Phenol Red Broth with Trehalose, Hardy Diagnostics
Catalog # 75814-676
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Phenol Red Broth with Trehalose, Hardy Diagnostics
Catalog # 75814-676
Supplier:  Hardy Diagnostics
Supplier Number:  Y317
CAS Number:  

Specifications

  • Description:
    Phenol Red Broth with Trehalose
  • Size:
    Pack of 20
  • Cat. No.:
    75814-676
  • Format:
    16×125 mm tube
  • Volume:
    10 ml
  • Supplier no.:
    Y317

Specifications

About this item

Phenol Red Broth with Trehalose is used for the differentiation of microorganisms by carbohydrate fermentation.

  • Contains a durham tube for gas detection
  • Differentiation of microorganisms by carbohydrate fermentation

For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.