Manganese Standards
About this item
Certified reference material for Manganese, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Certifications: ISO 17025/17034.
6 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Certified reference material for Manganese, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Certifications: ISO 17025/17034.
Product Family Options
Product Information
- SizeElementConcentrationColorMatrixPack typeAvailabilityPrice
- 125 mlManganese10000 µg/mlClearHNO₃ dil.Discontinued-Supplier Number: CGMN10-125MLSpecifications:MarketsourceMarketsource
- 500 mlManganese10000 µg/mlClearHNO₃ dil.Discontinued-Supplier Number: CGMN10-500MLSpecifications:MarketsourceMarketsource
- 500 mlManganese1000 µg/mlClearHNO₃Discontinued-Supplier Number: CGMN1-500MLSpecifications:MarketsourceMarketsource
- Supplier Number: CGMN10-125MLItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- Supplier Number: CGMN10-30MLItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- Supplier Number: CGMN10-500MLItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- Supplier Number: CGMN1-125MLSpecifications:New ProductNew Product
- Supplier Number: CGMN1-30MLSpecifications:New ProductNew Product
- Supplier Number: CGMN1-500MLItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product