Order Entry
Puerto Rico
ContactUsLinkComponent
Human LC-beta-Amyloid (1-42), Biotin, AnaSpec
Human LC-beta-Amyloid (1-42), Biotin, AnaSpec
Supplier:  Anaspec
Certificates
Human LC-beta-Amyloid (1-42), Biotin, AnaSpec
Supplier:  Anaspec

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Biotin
  • Species:
    Human
  • Storage Conditions:
    Upon receipt, store lyophilized powder at minus 20 degree celsius or lower. Reconstituted peptide should be aliquoted into several freezer vials and stored at minus 20 degree celsius or lower. Do not freeze thaw.
  • Protein Synonyms:
    amyloid beta (1-42) LC biotin|abeta (1-42) LC biotin|beta amyloid (1-42) LC biotin|amyloid 1-42 biotin LC
  • Protein/Peptide Name:
    LC-beta-Amyloid (1-42)
  • Purity:
    Greater than 95 % Purity by Peak area HPLC
  • Molecular Weight:
    4853.6 kDa
  • Sequence:
    Biotin-LC-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
  • Shipping Temperature:
    ambient

Specifications

Related Information

Frequently Bought Together