This peptide is beta-amyloid (1-42) with substitution of Ser26 to Cys. This peptide has been used in a number of fluorescent tagged experiments and suited for fluorescent probe labeling.
Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGCNKGAIIGLMVGGVVIA
MW: 4530.2 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Protein Synonyms:abeta (1-42) S26C|beta amyloid (1-42) S26C|amyloid beta (1-42) S26C|amyloid 1-42 S26C
- Protein/Peptide Name:[Cys26]-beta-Amyloid (1-42)
- Purity:95%
- Molecular Weight:4530.2 Da
- Sequence:DAEFRHDSGYEVHHQKLVFFAEDVGCNKGAIIGLMVGGVVIA
- Shipping Temperature:ambient