Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:HiLyte Fluor® 555
- Species:Human
- Size:0.1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:beta amyloid (1-42) HiLyte™ Fluor 555|abeta (1-42) HiLyte™ Fluor 555|amyloid 1-42 HiLyte™ Fluor 555|amyloid beta (1-42) HiLyte™ Fluor 555
- Protein/Peptide Name:beta-Amyloid (1-42)
- Purity:95%
- Molecular Weight:5366.57 Da
- Sequence:HiLyte™ Fluor 555-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
- Shipping Temperature:ambient
- Cat. No.:103003-170
- Supplier no.:AS-60480-01
Specifications
About this item
This is a fluorescent (HiLyte™ Fluor 555)-labeled ß-Amyloid peptide, Abs/Em=551/567 nm.
Sequence: HiLyte™ Fluor 555-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 5366.57 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citations:
Condello, C. et al. (2015). Microglia constitute a barrier that prevents neurotoxic protofibrillar Aβ42 hotspots around plaques. Nat Commun doi:10.1038/ncomms7176.
Epelbaum, S. et al. (2015). Acute amnestic encephalopathy in amyloid-ß oligomers injected mice is due to their widespread diffusion in vivo. Neurobiol Aging doi: 10.1016/j.neurobiolaging.2015.03.005.
Sonkar, VK. et al. (2014). Amyloid β peptide stimulates platelet activation through RhoA-dependent modulation of actomyosin organization. FASEB J 28, 1819. doi: 10.1096/fj.13-243691.