Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:TAMRA (5-Carboxytetramethylrhodamin)
- Species:Human
- Size:0.1 mg
- Storage Conditions:–20 °C
- Protein Synonyms:amyloid beta (1-42) TAMRA|abeta (1-42) TAMRA|beta amyloid (1-42) TAMRA|amyloid 1-42 TAMRA
- Protein/Peptide Name:beta-Amyloid (1-42)
- Purity:95%
- Molecular Weight:4926.6 Da
- Sequence:TAMRA-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
- Shipping Temperature:ambient
- Cat. No.:103003-166
- Supplier no.:AS-60476
This is a fluorescent (TAMRA)-labeled ß-Amyloid peptide, Abs/Em=544/572 nm.
Sequence: TAMRA-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 4926.6 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citation:
Ahn, HJ. et al. (2014). A novel Aβ-fibrinogen interaction inhibitor rescues altered thrombosis and cognitive decline in Alzheimer’s disease mice. J Exp Med 211, 1049. doi: 10.1084/jem.20131751.
Domert, J. et al. (2014). Spreading of amyloid-β peptides via neuritic cell-to-cell transfer is dependent on insufficient cellular clearance. Neurobiol Disease 65, 82. doi: 10.1016/j.nbd.2013.12.019.
Zhang, YH. et al. (2014). K114 Inhibits A-beta Aggregation and Inflammation In Vitro and In Vivo in AD/Tg Mice. Curr Alzheimer Res 11, 299.