Interleukin-12, IL-12, Human ELISA, High Sensitivity, BioVendor
Catalog # 102552-426
Supplier: BioVendor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Primary antibody reactivity:Human
- Target protein:IL-12
- Description:Interleukin-12 (IL-12) Human ELISA, High Sensitivity
- Size:1 kit
- Regulatory Status:RUO
- Cat. No.:102552-426
- Supplier no.:RGP016R
Specifications
About this item
- Calibration Range: 0.78–25 pg/mL
- Limit of Detection: 0.75 pg/mL
- Cell culture supernatant, Plasma, Serum: 100 uL/well
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.