ContactUsLinkComponent
Interleukin-10 Human ELISA, High Sensitivity, BioVendor
Interleukin-10 Human ELISA, High Sensitivity, BioVendor
Catalog # 102552-424
Supplier:  BioVendor
CAS Number:  
undefined
Interleukin-10 Human ELISA, High Sensitivity, BioVendor
Catalog # 102552-424
Supplier:  BioVendor
Supplier Number:  RGP015R
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Primary antibody reactivity:
    Human
  • Description:
    Interleukin-10 Human ELISA, High Sensitivity
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-424
  • Supplier no.:
    RGP015R
Specifications
About this item
IL-10 is an immunoregulatory protein produced by a number of cell lineages, including TH2 cells, B-cells and activated monocytes.
  • Calibration Range: 1.56–50 pg/mL
  • Limit of Detection: 1.3 pg/mL
  • Cell culture supernatant, Plasma, Serum: 100 uL/well
IL-10 is an immunoregulatory protein produced by a number of cell lineages, including TH2 cells, B-cells and activated monocytes. Il-10 is an important factor in understanding autoimmunity and as a target molecule in the development of anti-rejection therapies.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.