Order Entry
Puerto Rico
ContactUsLinkComponent
Trail Receptor 3, CD263, Human ELISA, BioVendor
Trail Receptor 3, CD263, Human ELISA, BioVendor
Catalog # 102552-632
Supplier:  BioVendor
CAS Number:  
undefined
Trail Receptor 3, CD263, Human ELISA, BioVendor
Catalog # 102552-632
Supplier:  BioVendor
Supplier Number:  RGP024R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    Trail Receptor 3, CD263
  • Description:
    Trail Receptor 3 (CD263) Human ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-632
  • Supplier no.:
    RGP024R

Specifications

About this item

Trail R3 antagonizes TRAIL-induced apoptosis.

  • Calibration Range: 312.5–10,000 pg/mL
  • Limit of Detection: 147 pg/mL
  • Cell culture lysate, Cell culture supernatant, Serum: 100 uL/well

Human Trail R1 (R2) is a TNF receptor , which is a receptor for Trail (APO2 ligand). Trail R1 and R2 are involved in apoptosis. In the Trail receptor family, Trail R1, but also Trail R2 transduce an apoptosis signal, whereas Trail R3 and Trail R4 antagonize TRAIL-induced apoptosis.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.