Order Entry
United States
ContactUsLinkComponent
Rat/Mouse FGF-21 ELISA, Biovendor
Rat/Mouse FGF-21 ELISA, Biovendor
Catalog # 102552-226
Supplier:  BioVendor
CAS Number:  
undefined
Rat/Mouse FGF-21 ELISA, Biovendor
Catalog # 102552-226
Supplier:  BioVendor
Supplier Number:  RD291108200R
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Rat
    Mouse
  • Target protein:
    FGF-21
  • Description:
    Rat/Mouse FGF-21 ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Sample Type:
    Serum
  • Detection Method:
    Colorimetric
  • Shelf Life:
    See expiration date
  • Detection Range:
    40 - 2560 pg/ml
  • Storage Temperature:
    2...8 °C
  • Sample Volume:
    Mouse samples: 7 µl/well
    Rat samples: 50 µl/well
  • Sensitivity:
    18.4 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-226
  • Supplier no.:
    RD291108200R
  • No. of tests:
    96 wells

Specifications

About this item

96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of FGF-21.

In the BioVendor Mouse and Rat FGF-21 ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with polyclonal anti-mouse FGF-21 antibody. After 60 minutes incubation and washing, biotin labelled polyclonal anti-mouse FGF-21 antibody is added and incubated for 60 minutes with captured FGF-21. After another washing, streptavidin HRP conjugate is added. After 30 minutes incubation and the last washing step, the remaining conjugate is allowed to react with the substrate solution (TMB).

The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of FGF-21. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.