Order Entry
United States
ContactUsLinkComponent
RBP4 Mouse/Rat ELISA, BioVendor
RBP4 Mouse/Rat ELISA, BioVendor
Catalog # 102552-688
Supplier:  BioVendor
CAS Number:  
undefined
RBP4 Mouse/Rat ELISA, BioVendor
Catalog # 102552-688
Supplier:  BioVendor
Supplier Number:  RAG007R
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    RBP4
  • Description:
    RBP4 Mouse/Rat ELISA
  • Size:
    1 kit
  • Environmentally Preferable:
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-688
  • Supplier no.:
    RAG007R

Specifications

About this item

Retinol binding protein (RBP) 4 is the only specific transport protein for vitamin A in the circulation whose function is to deliver vitamin to target tissues.

  • Calibration Range: 0.188-12 ng/mL
  • Limit of Detection: 60 pg/mL
  • Cell culture supernatant, Serum, Urine: sample diluiton will need to be determined, start at 10,000x dilution for serum and 10x for urine

Retinol binding protein (RBP) 4 is the only specific transport protein for vitamin A in the circulation whose function is to deliver vitamin to target tissues (1). In obesity and type 2 diabetes, expression of Glut4 is significantly impaired in adipocytes. Glucose transport via Glut4 is the rate-limiting step for glucose use by muscle and adipose tissue (2). Yang et al. noted that adipocyte-specific deletion of Gluts led to notable elevation of RBP4 causing systemic insulin resistance, and that reduction of RBP4 improved insulin resistance (3). This identified a novel role of RBP4 in regulating insulin action and RBP4 is recorded as an adipocyte-derived hormone. Thus, measurement of serum or plasma RBP4 is a useful means for understanding of metabolic disorders.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.