Specifications
- Target protein:IL-8 (CXCL8)
- Description:Interleukin-8 (IL-8) Monkey ELISA
- Size:1 kit
- Regulatory Status:RUO
- Cat. No.:102552-400
- Supplier no.:RAF075R
Specifications
About this item
IL-8 (8 kDa, 72 amino acids) is a non-glycosylated protein and it is produced by the processing of a precursor protein of 99 amino acids.
- Calibration Range: 19.5-1250 U/mL
- Limit of Detection: 2.5 U/mL
- Baboon serum, Cynomolgus serum: 50 uL/well
IL-8 (8 kDa, 72 amino acids) is a non-glycosylated protein and it is produced by the processing of a precursor protein of 99 amino acids. IL-8 is present in the synovial fluids of patients with rheumatoid arthritis and has been purified from plaque psoriasis scales. It is associated with sepsis, asthma and glomerulonephritis.