Order Entry
Puerto Rico
ContactUsLinkComponent
Interleukin-17F, IL-17F, Human ELISA, BioVendor
Interleukin-17F, IL-17F, Human ELISA, BioVendor
Catalog # 102552-332
Supplier:  BioVendor
CAS Number:  
undefined
Interleukin-17F, IL-17F, Human ELISA, BioVendor
Catalog # 102552-332
Supplier:  BioVendor
Supplier Number:  RAF043R
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-17F
  • Description:
    Interleukin-17F (IL-17F) Human ELISA
  • Size:
    1 kit
  • Regulatory Status:
    RUO
  • Cat. No.:
    102552-332
  • Supplier no.:
    RAF043R

Specifications

About this item

Interleukin-17 is involved in the regulation of hematopoiesis and its effects are dependent on the physiological/pathological status of the organism.

  • Calibration Range: 31.3-2000 pg/mL
  • Limit of Detection: 15.5 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Plasma-EDTA+PHMB, Plasma-Heparin, Serum: 50 uL/well

Animal studies, Cytokines and chemokines and related molecules

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.