Littmann® Lightweight II S.E Stethoscope is an ideal entry-level instrument, with a tunable diaphragm, dual-sided chestpiece, and reliable acoustic performance. The teardrop shape simplifies auscultation around blood pressure cuffs.
- Tunable diaphragm: Hear high or low frequency sounds by slightly adjusting pressure on the chestpiece
- Soft-sealing eartips provide an excellent acoustic seal and comfortable fit
- Lightest weight of all Littmann adult stethoscopes at just 118 grams
- Two-year warranty included